Gene Transcripts

Gene Symbol
Yipf6
Entrez Gene ID
77929
Gene Name
Yip1 domain family, member 6
Gene Aliases
A430107J06Rik
Chromosome Location
Chr. X: 98936321 - 98949021 on Build GRCm38
UniGene ID
Mm.212290
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207633.2 NP_997516.1 7 2403
GenBank mRNA AK030473.1 - 7 2405
AK045453.1 - 7 2115
AK053562.1 - 7 2402
AK143152.1 - 7 1972

View Full Product Details