Gene Transcripts

Gene Symbol
MCF2
Entrez Gene ID
4168
Gene Name
MCF.2 cell line derived transforming sequence
Gene Aliases
ARHGEF21, DBL
Chromosome Location
Chr. X: 139581768 - 139708279 on Build GRCh38
UniGene ID
Hs.387262
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB085901.1 - 2 155
AB085902.1 - 2 155
X12556.1 - 2 329

View Full Product Details