Gene Transcripts

Gene Symbol
Vprbp
Entrez Gene ID
321006
Gene Name
Vpr (HIV-1) binding protein
Gene Aliases
AI447437, B930007L02Rik, mKIAA0800
Chromosome Location
Chr. 9: 106814172 - 106880992 on Build GRCm38
UniGene ID
Mm.440089
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015507.2 NP_001015507.1 6 589
GenBank mRNA AK122372.1 - 6 552
BC065119.1 - 6 589 6853519

View Full Product Details