Gene Transcripts

Gene Symbol
Tmem86b
Entrez Gene ID
68255
Gene Name
transmembrane protein 86B
Gene Aliases
C330014O21Rik
Chromosome Location
Chr. 7: 4628040 - 4631395 on Build GRCm38
UniGene ID
Mm.389784
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023440.2 NP_075929.1 2 456
GenBank mRNA BC100527.1 - 2 456

View Full Product Details