Gene Transcripts

Gene Symbol
Ptprcap
Entrez Gene ID
499300
Gene Name
protein tyrosine phosphatase, receptor type, C-associated protein
Gene Aliases
Chromosome Location
Chr. 1: 219403970 - 219406129 on Build Rnor_6.0
UniGene ID
Rn.4188
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024289.1 NP_001019460.1 2 721
GenBank mRNA BC088279.1 - 2 721

View Full Product Details