Gene Transcripts
- Gene Symbol
- Klra5
- Entrez Gene ID
- 297666
- Gene Name
- killer cell lectin-like receptor, subfamily A, member 5
- Gene Aliases
- Ly-49.9
- Chromosome Location
- Chr. 4: 162657771 - 162682881 on Build Rnor_6.0
- UniGene ID
- Rn.37467
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_198746.1 |
NP_942041.1
|
|
1156 |
|
GenBank mRNA |
U56863.1 |
- |
|
1156 |
|
View Full Product Details