Gene Transcripts
- Gene Symbol
- H2-T22
- Entrez Gene ID
- 15039
- Gene Name
- histocompatibility 2, T region locus 22
- Gene Aliases
- 27b, H-2T17, H-2T22, H2-T17, T22b
- Chromosome Location
- Chr. 17: 36037133 - 36042742 on Build GRCm38
- UniGene ID
- Mm.195061
- Species
- Mus musculus
- Gene Symbol
- H2-T10
- Entrez Gene ID
- 15024
- Gene Name
- histocompatibility 2, T region locus 10
- Gene Aliases
- H-2T10
- Chromosome Location
- Chr. 17: 36115871 - 36121444 on Build GRCm38
- UniGene ID
- Mm.426031
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M35244.1 |
- |
8 |
1350 |
|
View Full Product Details