Gene Transcripts

Gene Symbol
H1fx
Entrez Gene ID
243529
Gene Name
H1 histone family, member X
Gene Aliases
Gm461, H1.10, H1X
Chromosome Location
Chr. 6: 87980421 - 87981482 on Build GRCm38
UniGene ID
Mm.33796
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198622.1 NP_941024.1 1 844
GenBank mRNA BC048709.1 - 1 844

View Full Product Details