Gene Transcripts

Gene Symbol
FUT5
Entrez Gene ID
2527
Gene Name
fucosyltransferase 5
Gene Aliases
FUC-TV
Chromosome Location
Chr. 19: 5865826 - 5870540 on Build GRCh38
UniGene ID
Hs.631843
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002034.2 NP_002025.2 2 1775
GenBank mRNA U27329.1 - 2 1776
U27330.1 - 2 1776

View Full Product Details