Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006342.2 | NP_006333.1 | 3 | 366 | |
XM_005247929.2 | XP_005247986.1 | 3 | 364 | ||
XM_005247930.2 | XP_005247987.1 | 3 | 473 | ||
XM_011513386.2 | XP_011511688.1 | 3 | 364 | ||
XM_017007653.1 | XP_016863142.1 | 3 | 418 | ||
GenBank mRNA | AF093543.1 | - | 3 | 319 | |
AF479620.1 | - | 3 | 311 | ||
AJ243997.1 | - | 3 | 311 | ||
AK225631.1 | - | 3 | 334 | ||
AK302463.1 | - | 3 | 337 | ||
AK304579.1 | - | 2 | 95 | ||
AK310660.1 | - | 2 | 89 | ||
BC106071.1 | - | 3 | 327 | 6148176 | |
BC111771.1 | - | 3 | 279 | 40034847 | |
DB098941.1 | - | 3 | 364 |