Gene Transcripts

Gene Symbol
CHAC1
Entrez Gene ID
79094
Gene Name
ChaC glutathione specific gamma-glutamylcyclotransferase 1
Gene Aliases
Chromosome Location
Chr. 15: 40942135 - 40956519 on Build GRCh38
UniGene ID
Hs.155569
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142776.1 NP_001136248.1 4 807
NM_024111.3 NP_077016.2 3 942
XM_017022563.1 XP_016878052.1 4 5466
XM_017022564.1 XP_016878053.1 5 3248
XM_017022565.1 XP_016878054.1 4 5455
GenBank mRNA BC001683.1 - 3 896
BC001847.2 - 3 887 2964714
BC019625.1 - 3 898 4473135
BM784841.1 - 1 268

View Full Product Details