Gene Transcripts
- Gene Symbol
- SLC30A9
- Entrez Gene ID
- 10463
- Gene Name
- solute carrier family 30 member 9
- Gene Aliases
- C4orf1, GAC63, HUEL, ZNT9
- Chromosome Location
- Chr. 4: 41990472 - 42087534 on Build GRCh38
- UniGene ID
- Hs.479634
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006345.3 |
NP_006336.3
|
1, 2 |
262 |
|
XM_017007654.1 |
XP_016863143.1
|
1, 2 |
296 |
|
GenBank mRNA |
AF006621.2 |
- |
1, 2 |
236 |
|
AK225470.1 |
- |
1, 2 |
199 |
|
AK299917.1 |
- |
1, 2 |
238 |
|
AK312838.1 |
- |
1, 2 |
262 |
|
AY319413.1 |
- |
2, 3 |
141 |
|
AY594282.1 |
- |
1, 2 |
116 |
|
BC000240.1 |
- |
1, 2 |
223 |
|
BC007732.2 |
- |
1, 2 |
223 |
4109702 |
BC016949.1 |
- |
1, 2 |
241 |
3850521 |
BG323207.1 |
- |
|
262 |
|
KU178526.1 |
- |
1, 2 |
116 |
|
KU178527.1 |
- |
1, 2 |
116 |
|
View Full Product Details