Gene Transcripts

Gene Symbol
NEUROD1
Entrez Gene ID
4760
Gene Name
neuronal differentiation 1
Gene Aliases
BETA2, BHF-1, MODY6, NEUROD, bHLHa3
Chromosome Location
Chr. 2: 181676106 - 181680665 on Build GRCh38
UniGene ID
Hs.574626
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002500.4 NP_002491.2 2 480
GenBank mRNA AB593068.1 - 2 332
AB593069.1 - 2 334
AB593070.1 - 2 332
AB593071.1 - 2 332
AK313799.1 - 2 332
BC009046.1 - 2 301 3873419
BT019731.1 - 1 233
D82347.1 - 2 342
DA486317.1 - 2 470
DB472071.1 - 2 425
KU178230.1 - 1 233

View Full Product Details