Gene Transcripts

Gene Symbol
C11orf70
Entrez Gene ID
85016
Gene Name
chromosome 11 open reading frame 70
Gene Aliases
Chromosome Location
Chr. 11: 102047409 - 102084560 on Build GRCh38
UniGene ID
Hs.98328
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032930.2 NP_116319.2 5, 6 638
GenBank mRNA AK094851.1 - 5, 6 618
BC006128.1 - 4, 5 842 3622924

View Full Product Details