Gene Transcripts
- Gene Symbol
- TSPAN18
- Entrez Gene ID
- 90139
- Gene Name
- tetraspanin 18
- Gene Aliases
- TSPAN
- Chromosome Location
- Chr. 11: 44727148 - 44932427 on Build GRCh38
- UniGene ID
- Hs.385634
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_130783.4 |
NP_570139.3
|
5, 6 |
573 |
|
XM_005253217.3 |
XP_005253274.1
|
7, 8 |
757 |
|
XM_006718372.3 |
XP_006718435.1
|
7, 8 |
844 |
|
XM_006718373.3 |
XP_006718436.1
|
6, 7 |
830 |
|
XM_011520459.2 |
XP_011518761.1
|
6, 7 |
720 |
|
XM_017018530.1 |
XP_016874019.1
|
6, 7 |
595 |
|
XM_017018531.1 |
XP_016874020.1
|
5, 6 |
517 |
|
GenBank mRNA |
AK027715.1 |
- |
4, 5 |
382 |
|
AK075547.1 |
- |
3, 4 |
306 |
|
AY358087.1 |
- |
4, 5 |
496 |
|
BC019342.2 |
- |
3, 4 |
345 |
3357734 |
DA734054.1 |
- |
6, 7 |
623 |
|
View Full Product Details