Gene Transcripts
- Gene Symbol
- EMID1
- Entrez Gene ID
- 129080
- Gene Name
- EMI domain containing 1
- Gene Aliases
- EMI5, EMU1
- Chromosome Location
- Chr. 22: 29205912 - 29259597 on Build GRCh38
- UniGene ID
- Hs.289106
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001267895.1 |
NP_001254824.1
|
1, 2 |
236 |
|
NM_133455.3 |
NP_597712.2
|
1, 2 |
236 |
|
XM_005261328.2 |
XP_005261385.1
|
1, 2 |
231 |
|
XM_005261329.2 |
XP_005261386.1
|
1, 2 |
225 |
|
XM_011529868.2 |
XP_011528170.1
|
1, 2 |
191 |
|
XM_011529869.2 |
XP_011528171.1
|
1, 2 |
190 |
|
XM_011529870.2 |
XP_011528172.1
|
1, 2 |
190 |
|
XM_011529871.2 |
XP_011528173.1
|
1, 2 |
193 |
|
XM_011529872.2 |
XP_011528174.1
|
1, 2 |
191 |
|
XM_011529873.2 |
XP_011528175.1
|
1, 2 |
191 |
|
GenBank mRNA |
AJ416090.1 |
- |
1, 2 |
109 |
|
BC046358.1 |
- |
1, 2 |
184 |
5770546 |
CR456407.1 |
- |
1, 2 |
119 |
|
View Full Product Details