Gene Transcripts

Gene Symbol
DEPDC7
Entrez Gene ID
91614
Gene Name
DEP domain containing 7
Gene Aliases
TR2, dJ85M6.4
Chromosome Location
Chr. 11: 33015864 - 33033582 on Build GRCh38
UniGene ID
Hs.280990
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139160.2 NP_631899.2 1 364
GenBank mRNA AF447588.1 - 1 364

View Full Product Details