Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166425.1 | NP_001159897.1 | 4 | 386 | |
NM_001282617.1 | NP_001269546.1 | 4 | 427 | ||
NM_001282618.1 | NP_001269547.1 | 3 | 458 | ||
NM_001282619.1 | NP_001269548.1 | 5 | 834 | ||
NM_001282620.1 | NP_001269549.1 | 4 | 710 | ||
NM_002070.3 | NP_002061.1 | 4 | 501 | ||
GenBank mRNA | AF493906.1 | - | 4 | 322 | |
AK055574.1 | - | 3 | 411 | ||
AK096299.1 | - | 4 | 603 | ||
AK096595.1 | - | 4 | 1105 | ||
AK126708.1 | - | 4 | 2138 | ||
AK131062.1 | - | 2 | 1149 | ||
AK302330.1 | - | 4 | 355 | ||
AK304473.1 | - | 4 | 266 | ||
AY677118.1 | - | 4 | 322 | ||
BC012138.1 | - | 4 | 482 | 4562585 | |
BC014627.2 | - | 4 | 439 | 2963802 | |
BC016995.1 | - | 5 | 785 | 4403631 | |
CR456783.1 | - | 4 | 322 | ||
DB044421.1 | - | 4 | 386 | ||
J03004.1 | - | 4 | 399 | ||
X04828.1 | - | 4 | 445 |