Gene Transcripts
- Gene Symbol
- ADGRG5
- Entrez Gene ID
- 221188
- Gene Name
- adhesion G protein-coupled receptor G5
- Gene Aliases
- GPR114, PGR27
- Chromosome Location
- Chr. 16: 57529339 - 57578182 on Build GRCh38
- UniGene ID
- Hs.187884
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001304376.2 |
NP_001291305.1
|
5, 6 |
540 |
|
NM_001318481.1 |
NP_001305410.1
|
5, 6 |
540 |
|
NM_153837.3 |
NP_722579.1
|
5, 6 |
1012 |
|
XM_011522949.2 |
XP_011521251.1
|
5, 6 |
556 |
|
XM_011522950.2 |
XP_011521252.1
|
5, 6 |
582 |
|
GenBank mRNA |
AK128298.1 |
- |
5, 6 |
691 |
|
AK160368.1 |
- |
5, 6 |
522 |
|
AK172765.1 |
- |
1, 2 |
29 |
|
AK292783.1 |
- |
5, 6 |
532 |
|
AK301357.1 |
- |
5, 6 |
460 |
|
AK303661.1 |
- |
5, 6 |
532 |
|
AY140956.1 |
- |
4, 5 |
437 |
|
AY358447.1 |
- |
5, 6 |
519 |
|
BC032401.1 |
- |
5, 6 |
960 |
5210813 |
View Full Product Details