Gene Transcripts

Gene Symbol
TRIM59
Entrez Gene ID
286827
Gene Name
tripartite motif containing 59
Gene Aliases
IFT80L, MRF1, RNF104, TRIM57, TSBF1
Chromosome Location
Chr. 3: 160435503 - 160449838 on Build GRCh38
UniGene ID
Hs.212957
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173084.2 NP_775107.1 3 1160
GenBank mRNA AK291284.1 - 3 1082
AY159379.1 - 3 1160
BC109259.1 - 3 1147 40025031
BC109260.1 - 3 1147 40025036

View Full Product Details