Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173551.4 | NP_775822.3 | 10 | 1891 | |
XM_005251793.4 | XP_005251850.1 | 10 | 1878 | ||
XM_005251794.4 | XP_005251851.1 | 10 | 1878 | ||
XM_006716998.3 | XP_006717061.1 | 10 | 1878 | ||
XM_006716999.2 | XP_006717062.1 | 10 | 2963 | ||
XM_011518357.2 | XP_011516659.1 | 10 | 3960 | ||
XM_017014445.1 | XP_016869934.1 | 10 | 1878 | ||
XM_017014446.1 | XP_016869935.1 | 9 | 3839 | ||
XM_017014447.1 | XP_016869936.1 | 6 | 672 | ||
GenBank mRNA | AK094247.1 | - | 3 | 245 | |
AK126614.1 | - | 8 | 945 | ||
AK127702.1 | - | 9 | 1478 | ||
BC064367.1 | - | 7 | 878 | 5562895 | |
CR749467.1 | - | 10 | 1562 | ||
CR749472.1 | - | 8 | 1242 |