Gene Transcripts

Gene Symbol
B4GALNT3
Entrez Gene ID
283358
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 3
Gene Aliases
Chromosome Location
Chr. 12: 460377 - 561892 on Build GRCh38
UniGene ID
Hs.504416
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173593.3 NP_775864.3 14 1847
GenBank mRNA AB089940.1 - 14 1847
AK093503.1 - 12 1572
AK097681.1 - 1 232
AK131277.1 - 1 1238

View Full Product Details