Gene Transcripts
- Gene Symbol
- SSNA1
- Entrez Gene ID
- 8636
- Gene Name
- SS nuclear autoantigen 1
- Gene Aliases
- N14, NA-14, NA14
- Chromosome Location
- Chr. 9: 137188602 - 137190370 on Build GRCh38
- UniGene ID
- Hs.530314
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
Z96932.1 |
- |
2, 3 |
306 |
|
View Full Product Details