Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013286.4 | NP_037418.3 | 1 | 2177 | |
GenBank mRNA | AL831838.1 | - | 1 | 1583 | |
AY545557.1 | - | 1 | 151 | ||
BC001367.2 | - | 1 | 720 | ||
BC139836.1 | - | 2 | 1388 | 40123447 | |
L13434.1 | - | 1 | 1934 |