Gene Transcripts

Gene Symbol
ANKRD11
Entrez Gene ID
29123
Gene Name
ankyrin repeat domain 11
Gene Aliases
ANCO-1, ANCO1, LZ16, T13
Chromosome Location
Chr. 16: 89267619 - 89490561 on Build GRCh38
UniGene ID
Hs.335003
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256182.1 NP_001243111.1 4, 5 615
NM_001256183.1 NP_001243112.1 3, 4 541
NM_013275.5 NP_037407.4 3, 4 544
XM_006721181.1 XP_006721244.1 2, 3 224
XM_011523051.2 XP_011521353.1 4, 5 604
XM_011523053.2 XP_011521355.1 4, 5 710
XM_011523054.2 XP_011521356.1 3, 4 527
XM_011523056.2 XP_011521358.1 3, 4 527
XM_011523057.2 XP_011521359.1 3, 4 527
XM_017023182.1 XP_016878671.1 3, 4 977
XM_017023183.1 XP_016878672.1 4, 5 713
XM_017023184.1 XP_016878673.1 4, 5 710
XM_017023185.1 XP_016878674.1 2, 3 351
XM_017023186.1 XP_016878675.1 6, 7 1087
XM_017023187.1 XP_016878676.1 2, 3 439
GenBank mRNA AB209186.1 - 2, 3 189
AF121775.1 - 3, 4 301
AK125549.1 - 2, 3 520
AK290994.1 - 2, 3 223
AY373756.1 - 3, 4 544
AY533563.1 - 4, 5 372
BC017437.1 - 3, 4 513
BC025283.1 - 2, 3 168
BC052290.1 - 3, 4 459
BC069013.1 - 3, 4 459
BQ957747.1 - 532
KJ534770.1 - 1, 2 83
KM670019.1 - 142
KM670020.1 - 142

View Full Product Details