Gene Transcripts
- Gene Symbol
- ARMC5
- Entrez Gene ID
- 79798
- Gene Name
- armadillo repeat containing 5
- Gene Aliases
- AIMAH2
- Chromosome Location
- Chr. 16: 31458273 - 31467167 on Build GRCh38
- UniGene ID
- Hs.732945
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001105247.1 |
NP_001098717.1
|
1, 2 |
997 |
|
NM_001288767.1 |
NP_001275696.1
|
3, 4 |
877 |
|
NM_001301820.1 |
NP_001288749.1
|
2, 3 |
1012 |
|
NM_024742.2 |
NP_079018.1
|
1, 2 |
997 |
|
XM_006721091.2 |
XP_006721154.1
|
2, 3 |
1130 |
|
GenBank mRNA |
AK024430.1 |
- |
1, 2 |
490 |
|
AK294897.1 |
- |
2, 3 |
919 |
|
AK295790.1 |
- |
3, 4 |
877 |
|
AK297115.1 |
- |
1, 2 |
32 |
|
AY217348.1 |
- |
1, 2 |
997 |
|
BC014945.1 |
- |
2, 3 |
402 |
4860377 |
View Full Product Details