Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020119.3 | NP_064504.2 | 2 | 818 | |
NM_024625.3 | NP_078901.3 | 2 | 818 | ||
XM_005250501.2 | XP_005250558.1 | 2 | 819 | ||
XM_011516412.2 | XP_011514714.1 | 2 | 819 | ||
GenBank mRNA | AK055851.1 | - | 2 | 453 | |
AK292811.1 | - | 2 | 486 | ||
BC025308.1 | - | 2 | 747 | 5418915 | |
BC033105.1 | - | 2 | 747 | 5418956 | |
BC040956.1 | - | 2 | 818 | 5531412 | |
BX571742.1 | - | 2 | 817 | ||
BX647974.1 | - | 2 | 454 |