Gene Transcripts

Gene Symbol
LMO1
Entrez Gene ID
4004
Gene Name
LIM domain only 1
Gene Aliases
RBTN1, RHOM1, TTG1
Chromosome Location
Chr. 11: 8224304 - 8268635 on Build GRCh38
UniGene ID
Hs.654426
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270428.1 NP_001257357.1 2 282
NM_002315.2 NP_002306.1 2 614
NR_073006.1 2 614
XM_006718228.3 XP_006718291.1 2 1660
XM_011520098.2 XP_011518400.1 2 149
XM_011520099.2 XP_011518401.1 2 166
GenBank mRNA BC039512.1 - 2 282
BC069673.1 - 2 156 7262648
BC069752.1 - 2 490 7262660
BC069793.1 - 2 490 7262577
BC096056.3 - 2 124 40000075
BC096057.3 - 2 123 40000076
BC096058.1 - 2 124 40000078
M26682.1 - 2 595
X64244.1 - 2 98
X64245.1 - 1 112

View Full Product Details