Gene Transcripts

Gene Symbol
PPP1R14A
Entrez Gene ID
94274
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14A
Gene Aliases
CPI-17, CPI17, PPP1INL
Chromosome Location
Chr. 19: 38251237 - 38256591 on Build GRCh38
UniGene ID
Hs.631569
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033256.2 NP_150281.1 2 506
GenBank mRNA AB056508.1 - 2 447
AF123763.1 - 2 297
AY050670.1 - 2 291
BC021089.1 - 2 271 5019808

View Full Product Details