Gene Transcripts

Gene Symbol
Cyp19a1
Entrez Gene ID
25147
Gene Name
cytochrome P450, family 19, subfamily a, polypeptide 1
Gene Aliases
Aromatase, Cyp19, Cyp19a, p450arom
Chromosome Location
Chr. 8: 58744849 - 58772408 on Build Rnor_6.0
UniGene ID
Rn.21402
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017085.2 NP_058781.2 3 896
GenBank mRNA EF091858.1 - 2 183

View Full Product Details