Gene Transcripts
- Gene Symbol
- Cyp19a1
- Entrez Gene ID
- 25147
- Gene Name
- cytochrome P450, family 19, subfamily a, polypeptide 1
- Gene Aliases
- Aromatase, Cyp19, Cyp19a, p450arom
- Chromosome Location
- Chr. 8: 58744849 - 58772408 on Build Rnor_6.0
- UniGene ID
- Rn.21402
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017085.2 |
NP_058781.2
|
6 |
1274 |
|
View Full Product Details