Gene Transcripts

Gene Symbol
Kcnj8
Entrez Gene ID
25472
Gene Name
potassium channel, inwardly rectifying subfamily J, member 8
Gene Aliases
Kir6.1, UKATP1, uKATP-1
Chromosome Location
Chr. 4: 176783287 - 176789143 on Build Rnor_6.0
UniGene ID
Rn.229203
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017099.4 NP_058795.3 3 831
GenBank mRNA AB043636.1 - 2 661
AB043637.1 - 2 661
BC078863.1 - 814 7128284
D42145.1 - 3 834

View Full Product Details