Gene Transcripts

Gene Symbol
5430421N21Rik
Entrez Gene ID
100126226
Gene Name
RIKEN cDNA 5430421N21 gene
Gene Aliases
Krt83
Chromosome Location
Chr. 15: 101485130 - 101491512 on Build GRCm38
UniGene ID
Mm.347934
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001201323.1 NP_001188252.1 9 1540
GenBank mRNA AK133718.1 - 9 1541
M92088.1 - 9 1525

View Full Product Details