Gene Transcripts

Gene Symbol
Gm14124
Entrez Gene ID
100216455
Gene Name
predicted gene 14124
Gene Aliases
EG640962, ENSMUSG00000074735, Gm10749, OTTMUSG00000015734
Chromosome Location
Chr. 2: 150257517 - 150270300 on Build GRCm38
UniGene ID
Mm.484816
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142410.1 NP_001135882.1 4 1891

View Full Product Details