Gene Transcripts

Gene Symbol
Gm20172
Entrez Gene ID
100504323
Gene Name
predicted gene, 20172
Gene Aliases
Chromosome Location
Chr. 1: 25276404 - 25277954 on Build GRCm38
UniGene ID
Mm.484176
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204913.1 NP_001191842.1 1 982
GenBank mRNA AK084043.1 - 1 982
AK148788.1 - 1 982

View Full Product Details