Gene Transcripts

Gene Symbol
H2-T22
Entrez Gene ID
15039
Gene Name
histocompatibility 2, T region locus 22
Gene Aliases
27b, H-2T17, H-2T22, H2-T17, T22b
Chromosome Location
Chr. 17: 36037133 - 36042742 on Build GRCm38
UniGene ID
Mm.195061
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010397.4 NP_034527.1 1 211
GenBank mRNA AK161744.1 - 1 210
M35243.1 - 1 180
M35245.1 - 1 211
M35247.1 - 1 192

View Full Product Details