Gene Transcripts

Gene Symbol
Sptan1
Entrez Gene ID
20740
Gene Name
spectrin alpha, non-erythrocytic 1
Gene Aliases
2610027H02Rik, Spna-2, Spna2
Chromosome Location
Chr. 2: 29965553 - 30031451 on Build GRCm38
UniGene ID
Mm.204969
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177667.1 NP_001171138.1 36 4911
NM_001177668.1 NP_001171139.1 35 4851
GenBank mRNA AK147317.1 - 35 4737
BC022120.1 - 5 604
BC027791.1 - 13 1343
BC150941.1 - 36 4747 9087852
BC158015.1 - 36 4747 9007321
BC172095.1 - 35 4687 9054381

View Full Product Details