Gene Transcripts
- Gene Symbol
- Golgb1
- Entrez Gene ID
- 224139
- Gene Name
- golgi autoantigen, golgin subfamily b, macrogolgin 1
- Gene Aliases
- 4930428L02Rik, 628101, 6330407A06Rik, AU042952, C130074L01Rik, F730017E11Rik, Gm6840, mKIAA4151
- Chromosome Location
- Chr. 16: 36875173 - 36933085 on Build GRCm38
- UniGene ID
- Mm.244815
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030035.1 |
NP_084311.1
|
13 |
4113 |
|
GenBank mRNA |
BC150731.1 |
- |
13 |
4090 |
|
View Full Product Details