Gene Transcripts

Gene Symbol
A230065H16Rik
Entrez Gene ID
380787
Gene Name
RIKEN cDNA A230065H16 gene
Gene Aliases
Gm898, Lbh2
Chromosome Location
Chr. 12: 111402047 - 111412077 on Build GRCm38
UniGene ID
Mm.125602
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101503.1 NP_001094973.1 4 333
GenBank mRNA AK038816.1 - 4 333

View Full Product Details