Gene Transcripts

Gene Symbol
Vmn2r50
Entrez Gene ID
434117
Gene Name
vomeronasal 2, receptor 50
Gene Aliases
EG434117
Chromosome Location
Chr. 7: 10037235 - 10053178 on Build GRCm38
UniGene ID
Mm.247383
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105178.1 NP_001098648.1 6 2353

View Full Product Details