Gene Transcripts

Gene Symbol
4933403O08Rik
Entrez Gene ID
71030
Gene Name
RIKEN cDNA 4933403O08 gene
Gene Aliases
Chromosome Location
Chr. X: 112171438 - 112249350 on Build GRCm38
UniGene ID
Mm.158538
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177389.1 NP_001170860.1 2 968
GenBank mRNA AK016644.1 - 2 965
BC115851.1 - 2 520
BC127170.1 - 1 856

View Full Product Details