Gene Transcripts

Gene Symbol
Ccdc88b
Entrez Gene ID
78317
Gene Name
coiled-coil domain containing 88B
Gene Aliases
2610041P08Rik, Ccdc88
Chromosome Location
Chr. 19: 6844623 - 6858328 on Build GRCm38
UniGene ID
Mm.329596
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081291.1 NP_001074760.1 27 4928
GenBank mRNA AK019167.1 - 1 276
BC038890.1 - 1 472
BC076600.1 - 18 3812

View Full Product Details