Gene Transcripts

Gene Symbol
Tgm2
Entrez Gene ID
56083
Gene Name
transglutaminase 2
Gene Aliases
TgaseII
Chromosome Location
Chr. 3: 154597165 - 154627257 on Build Rnor_6.0
UniGene ID
Rn.10
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019386.2 NP_062259.2 13 2092
GenBank mRNA AF106325.1 - 12 2043
BC062062.1 - 13 2092 5599202

View Full Product Details