Gene Transcripts
- Gene Symbol
- Grid1
- Entrez Gene ID
- 79219
- Gene Name
- glutamate receptor, ionotropic, delta 1
- Gene Aliases
- GluD1
- Chromosome Location
- Chr. 16: 11170831 - 11932197 on Build Rnor_6.0
- UniGene ID
- Rn.10040
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024378.1 |
NP_077354.1
|
2 |
351 |
|
GenBank mRNA |
U08255.1 |
- |
2 |
351 |
|
View Full Product Details