Gene Transcripts
- Gene Symbol
- Pax4
- Entrez Gene ID
- 83630
- Gene Name
- paired box 4
- Gene Aliases
- Chromosome Location
- Chr. 4: 55735643 - 55753635 on Build Rnor_6.0
- UniGene ID
- Rn.14531
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031799.1 |
NP_113987.1
|
2, 3 |
200 |
|
XM_006236231.2 |
XP_006236293.1
|
3, 4 |
6253 |
|
XM_006236232.2 |
XP_006236294.1
|
4, 5 |
1249 |
|
XM_006236233.2 |
XP_006236295.1
|
4, 5 |
691 |
|
GenBank mRNA |
AF053100.1 |
- |
2, 3 |
200 |
|
AF053101.1 |
- |
2, 3 |
200 |
|
AF053102.1 |
- |
2, 3 |
200 |
|
AF053103.1 |
- |
2, 3 |
200 |
|
AF198155.1 |
- |
1, 2 |
152 |
|
AF198156.1 |
- |
1, 2 |
152 |
|
View Full Product Details