Gene Transcripts
- Gene Symbol
- Cds2
- Entrez Gene ID
- 114101
- Gene Name
- CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 2
- Gene Aliases
- Chromosome Location
- Chr. 3: 124896657 - 124935221 on Build Rnor_6.0
- UniGene ID
- Rn.212746
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053643.1 |
NP_446095.1
|
11 |
1076 |
|
GenBank mRNA |
AB052898.1 |
- |
11 |
1076 |
|
View Full Product Details