Gene Transcripts

Gene Symbol
Atp2c1
Entrez Gene ID
170699
Gene Name
ATPase, Ca++ transporting, type 2C, member 1
Gene Aliases
Spca1
Chromosome Location
Chr. 8: 113953959 - 114039965 on Build Rnor_6.0
UniGene ID
Rn.5805
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_131907.2 NP_571982.2 3, 4 592
GenBank mRNA BC086994.1 - 3, 4 592
M93017.1 - 3, 4 303

View Full Product Details