Gene Transcripts

Gene Symbol
Adra1b
Entrez Gene ID
11548
Gene Name
adrenergic receptor, alpha 1b
Gene Aliases
[a]1b
Chromosome Location
Chr. 11: 43774605 - 43901251 on Build GRCm38
UniGene ID
Mm.39086
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284380.1 NP_001271309.1 3 1142
NM_001284381.1 NP_001271310.1 4 1292
NM_007416.4 NP_031442.2 1 1138
GenBank mRNA AK004891.1 - 4 1249
AK043877.1 - 1 983
AK139448.1 - 1 1141
BC018330.1 - 3 1115
BC037002.1 - 3 1116
BC138653.1 - 1 856 8861673
BC138654.1 - 1 856 8861674
S80219.1 - 1 25
Y12738.1 - 1 1457

View Full Product Details