Gene Transcripts

Gene Symbol
ZSCAN16
Entrez Gene ID
80345
Gene Name
zinc finger and SCAN domain containing 16
Gene Aliases
ZNF392, ZNF435, dJ265C24.3
Chromosome Location
Chr. 6: 28123753 - 28130086 on Build GRCh38
UniGene ID
Hs.656641
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320555.1 NP_001307484.1 2 318
NM_001320556.1 NP_001307485.1 2 268
NM_001320557.1 NP_001307486.1 2 268
NM_001320558.1 NP_001307487.1 2 268
NM_025231.2 NP_079507.1 2 268
XM_017011324.1 XP_016866813.1 2 1237
GenBank mRNA AK025844.1 - 2 215
AK294017.1 - 2 215
AK309837.1 - 2 215
BC004255.1 - 2 189 3609511
CB158915.1 - 2 197
CX865974.1 - 1 158
DA093436.1 - 2 224
DB158261.1 - 2 268

View Full Product Details