Gene Transcripts

Gene Symbol
Mak
Entrez Gene ID
17152
Gene Name
male germ cell-associated kinase
Gene Aliases
A930010O05Rik
Chromosome Location
Chr. 13: 41025007 - 41079706 on Build GRCm38
UniGene ID
Mm.8149
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145802.1 NP_001139274.1 2, 3 442
NM_001145803.1 NP_001139275.1 2, 3 442
NM_008547.2 NP_032573.2 1, 2 337
GenBank mRNA BC050009.1 - 2, 3 381
BQ930470.1 - 2, 3 357
X66983.1 - 1, 2 331

View Full Product Details